EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Esters of 2-furanacrylic acid.
PMID 20292450
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F20807
3-(2-Furyl)acrylic acid, 99%
C7H6O3