EMAIL THIS PAGE TO A FRIEND
The clinical respiratory journal
Vitamin and mineral status in elderly patients with chronic obstructive pulmonary disease.
PMID 20298274
Abstract
Eating problems are common in patients with chronic obstructive pulmonary disease (COPD), and intake of micronutrients might be lower than recommended. To study dietary intake, serum and urinary concentration of trace elements and vitamins in elderly underweight patients with established severe COPD. Outpatients at a university clinic for lung medicine, with COPD, 70-85 years old, with no other serious disease, and with a body mass index (BMI) of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
136441
Dimethyl malonate, 98%
C5H8O4
63380
Dimethyl malonate, purum, ≥96.0% (GC)
C5H8O4