EMAIL THIS PAGE TO A FRIEND
Talanta
Flow-injection catalytic spectrophotometic determination of molybdenum(VI) in plants using bromate oxidative coupling of p-hydrazinobensenesulfonic acid with N-(1-naphthyl)ethylenediamine.
PMID 20298854
Abstract
A novel flow-injection spectrophotometry has been developed for the determination of molybdenum(VI) at nanograms per milliliter levels. The method is based on the catalytic effect of molybdenum(VI) on the bromate oxidative coupling of p-hydrazinobenzenesulfonic acid with N-(1-naphthyl)ethylenediamine to form an azo dye (lambda(max)=530nm). Chromotropic acid (4,5-dihydroxy-2,7-naphthalenedisulfonic acid) acted as an effective activator for the molybdenum(VI)-catalyzed reaction and increased the sensitivity of the method. The reaction was monitored by measuring the change in absorbance of the dye produced. The proposed method allowed the determination of molybdenum(VI) in the range 1.0-20ngmL(-1) with sample throughput of 15h(-1). The limit of detection was 0.5ngmL(-1) and a relative standard deviation for 10ngmL(-1) molybdenum(VI) (n=10) was 2.5%. The interfering ions were eliminated by using the combination of a masking agent and on-line minicolumn packed with cation exchanger. The present method was successfully applied to the determination of molybdenum(VI) in plant foodstuffs.