EMAIL THIS PAGE TO A FRIEND
Anesthesia and analgesia
Effect on postoperative sore throat of spraying the endotracheal tube cuff with benzydamine hydrochloride, 10% lidocaine, and 2% lidocaine.
PMID 20304980
Abstract
Postoperative sore throat (POST) is a common complication after endotracheal intubation. We compared the effectiveness on POST of spraying the endotracheal tube (ETT) cuff with benzydamine hydrochloride, 10% lidocaine, and 2% lidocaine. Three hundred seventy-two patients were randomly allocated into 4 groups. The ETT cuffs in each group were sprayed with benzydamine hydrochloride, 10% lidocaine hydrochloride, 2% lidocaine hydrochloride, or normal saline before endotracheal intubation. After insertion, the cuffs were inflated to an airway leak pressure of 20 cm H(2)O. Anesthesia was maintained with propofol. The patients were examined for sore throat (none, mild, moderate, or severe) at 1, 6, 12, and 24 hours after extubation. The highest incidence of POST occurred at 6 hours after extubation in all groups. There was a significantly lower incidence of POST in the benzydamine group than 10% lidocaine, 2% lidocaine, and normal saline groups (P < 0.05) at each observation time point. At 6 hours after extubation, the incidence of POST was significantly lower in the benzydamine group (17.0%) compared with 10% lidocaine (53.7%), 2% lidocaine (37.0%), and normal saline (40.8%) groups (P < 0.05). The benzydamine group had significantly decreased severity of POST compared with the 10% lidocaine, 2% lidocaine, and normal saline groups (P < 0.05) at each observation time point. Compared with the 2% lidocaine and normal saline groups, the 10% lidocaine group had significantly increased severity of POST at 1, 6, and 12 hours after extubation. There were no significant differences among groups in local or systemic side effects. Spraying benzydamine hydrochloride on the ETT cuff is a simple and effective method to reduce the incidence and severity of POST.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5524
Benzydamine hydrochloride, analytical standard
C19H23N3O · HCl