EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
A class of 3S-2-aminoacyltetrahydro-beta-carboline-3-carboxylic acids: their facile synthesis, inhibition for platelet activation, and high in vivo anti-thrombotic potency.
PMID 20329729
Abstract
3S-Tetrahydro-beta-carboline-3-carboxylic acid (TCCA) effectively inhibits ADP-induced platelet activation. This paper used TCCA as a lead, modified its 2-position with amino acids, and provided 20 novel 3S-2-aminoacyl-1,2,3,4-tetrahydro-beta-carboline-3-carboxylic acids (5a-t). With the in vitro assay, it was demonstrated that this modification diminished the IC(50) values from 701 nM of TCCA to 10 nM of 5a-t. With the in vivo assay, it was demonstrated that this modification reduced the efficacious dose from 5.0 micromol/kg of TCCA to 0.1 micromol/kg of 5a-t. Comparing the Cerius based conformation of them with that of their analogues, the 3-position modified TCCA, it was suggested that the comparatively unfolded conformation was one of the important factors of enhancing the in vivo antithrombotic potency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
103276
Harmane, 98%
C12H10N2
N6252
Norharmane, crystalline
C11H8N2