EMAIL THIS PAGE TO A FRIEND
Natural product communications
Secondary metabolites of the phytopathogen Peronophythora litchii.
PMID 20334136
Abstract
Two new metabolites, 4R*-hydroxy-3,5R*-dimethoxycyclohex-2-enone (1) and 2-hydroxy-N-(2-hydroxyphenyl)propionamide (2), were isolated from solid cultures of the phytopathogen Peronophythora litchii, along with 4-hydroxybenzyl alcohol (3) and methyl D-indole-3-lactate (4). Their structures were elucidated on the basis of spectroscopic and spectrometric data. These metabolites were not toxic to brine shrimps (LC50 > 500 microg/mL).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H20806
4-Hydroxybenzyl alcohol, 99%
C7H8O2
W398705
4-Hydroxybenzyl alcohol, ≥98%, FG
C7H8O2