EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Thermal [4 + 2] cycloadditions of 3-acetyl-, 3-carbamoyl-, and 3-ethoxycarbonyl-coumarins with 2,3-dimethyl-1,3-butadiene under solventless conditions: a structural study.
PMID 20335998
Abstract
The thermal [4+2] cycloadditions of 3-acetyl-, 3-carbamoyl, and 3-ethoxycarbonylcoumarins with 2,3-dimethyl-1,3-butadiene under solvent free conditions are reported, as well as the epoxidation reactions of some adducts. Discussion is focused on the structural features of the Diels-Alder adducts and their epoxides, based upon NMR, X-ray, and mass spectral data, and supported by ab initio theoretical calculations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
145491
2,3-Dimethyl-1,3-butadiene, 98%, contains 100 ppm BHT as stabilizer
C6H10