EMAIL THIS PAGE TO A FRIEND
Folia microbiologica
Benomyl-resistant mutant strain of Trichoderma sp. with increased mycoparasitic activity.
PMID 20336512
Abstract
Application of UV radiation to the strain Trichoderma sp. T-bt (isolated from lignite) resulted in the T-brm mutant which was resistant to the systemic fungicide benomyl. The tub2 gene sequence in the T-brm mutant differed from the parent as well as the collection strain (replacing tyrosine with histidine in the TUB2 protein). Under in vitro conditions this mutant exhibited a higher mycoparasitic activity toward phytopathogenic fungi.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45339
Benomyl, PESTANAL®, analytical standard
C14H18N4O3