EMAIL THIS PAGE TO A FRIEND
Turkiye parazitolojii dergisi
[Chewing-lice species (Phthiraptera) found on domestic and wild birds in Turkey].
PMID 20340090
Abstract
This study was carried out 48 domestic and wild bird samples belonging to 15 different species. Birds that were killed by traffic on the roads as well as birds that were ill or injured were brought for investigation to the Veterinary Faculty or Animal Keeping House of Konya Municipal. Firstly, all of them were inspected macroscopically for ectoparasites. Then, they were treated with an insecticidal drug, propoxur in a cartoon box. One long-eared owl, one Eurasian buzzard, four long-legged buzzards and five rock pigeons were found to be infested with lice, the others were not. The lice were mounted on slides into Canada balsam after being cleared in KOH 10 % and were identified to species under the light microscope. The lice were identified as Strigiphilus barbatus collected from the long-eared owl, as Kurodaia fulvofasciata collected from the Eurasian buzzard, as Crasspedorrhynchus platystomus, Degeeriella fulva, Colpocephalum nanum collected from long-legged buzzards and as Columbicola columbae collected from rock pigeons. This is the first time that Kurodaia fulvofasciata from the Eurasian buzzard and Strigiphilus barbatus from the long-eared owl has been recorded in Turkey.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45644
Propoxur, PESTANAL®, analytical standard
C11H15NO3