EMAIL THIS PAGE TO A FRIEND
Synapse (New York, N.Y.)
Orofacial movements in phospholipase C-related catalytically inactive protein-1/2 double knockout mice: Effect of the GABAergic agent diazepam and the D(1) dopamine receptor agonist SKF 83959.
PMID 20340178
Abstract
Orofacial movements are regulated by D(1)-like dopamine receptors interacting with additional mechanisms. Phospholipase C-related catalytically inactive protein (PRIP) regulates cell surface expression of GABA(A) receptors containing a gamma2 subunit. Mutant mice with double knockout of PRIP-1 and PRIP-2 were used to investigate aspects of GABAergic regulation of orofacial movements and interactions with D(1) mechanisms. Vertical jaw movements, tongue protrusions and movements of the head and vibrissae were reduced in PRIP-1/2 double knockouts. The GABA(A)ergic agent diazepam reduced movements of the head and vibrissae; these effects were unaltered in PRIP-1/2 double knockouts. The D(1)-like agonist SKF 83959 induced vertical jaw movements, incisor chattering, and movements of the head and vibrissae that were unaltered in PRIP-1/2 double knockouts. However, SKF 83959-induced tongue protrusions were reduced in PRIP-1/2 double knockouts. PRIP-mediated regulation of GABA(A)ergic receptor mechanisms influences topographically distinct aspects of orofacial movement and interacts with D(1) receptor systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S2816
SKF-83959 hydrobromide, ≥98% (HPLC), solid
C18H20NClO2 · HBr