EMAIL THIS PAGE TO A FRIEND
Molecular Ecology
Protein expression and genetic structure of the coral Porites lobata in an environmentally extreme Samoan back reef: does host genotype limit phenotypic plasticity?
PMID 20345691