EMAIL THIS PAGE TO A FRIEND
BJOG : an international journal of obstetrics and gynaecology
A randomised controlled trial of 300 versus 225 IU recombinant FSH for ovarian stimulation in predicted normal responders by antral follicle count.
PMID 20353458
Abstract
To test the hypothesis that among women predicted to have a normal ovarian response, ovarian stimulation using 300 IU follicle-stimulating hormone (FSH) results in the retrieval of more mature oocytes than 225 IU during in vitro fertilisation (IVF)/intracytoplasmic sperm injection (ICSI) treatment. Prospective randomised controlled study. University-based assisted conception unit. A cohort of 131 women predicted to have a normal ovarian response to gonadotrophin stimulation, based on antral follicle count. Subjects undergoing their first cycle of IVF/ICSI were randomised to receive a fixed daily dose of 300 (experimental arm) or 225 IU (control arm) of recombinant FSH (Gonal-F). Number of mature oocytes retrieved and live birth rates. The number (mean +/- standard deviation) of mature oocytes retrieved (8.2 +/- 5.0 versus 9.0 +/- 4.8, for 300 and 225 IU, respectively; P = 0.34) was similar in each group. There were no differences between the 300- and 225 IU arms in live birth rates (31 versus 41%, respectively; P = 0.25), cycle cancellations resulting from insufficient ovarian response (0 versus 6.1%, respectively; P = 0.12), and prevalence of moderate (3.1 versus 3.0, respectively; P = 1.0) and severe (0 versus 1.5%, respectively; P = 1.0) ovarian hyperstimulation syndrome. The use of a higher daily dose of 300 IU of recombinant FSH for ovarian stimulation does not improve the number of mature oocytes retrieved, or live birth rates, among women with a predicted normal response during conventional IVF/ICSI.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7662
Nafarelin acetate salt hydrate, ≥95% (HPLC), solid
C66H83N17O13 · xC2H4O2 · yH2O