EMAIL THIS PAGE TO A FRIEND
Cellular and molecular life sciences : CMLS
Lysosomal chymotrypsin B potentiates apoptosis via cleavage of Bid.
PMID 20361227
Abstract
We have reported that chymotrypsin B (CtrB) is not just a digestive enzyme but is also stored in lysosomes. Herein, we demonstrated a broad distribution of CtrB and explored the involvement of CtrB in apoptosis. Exposure of RH-35 cells to H(2)O(2) or palmitate induced the redistribution of lysosomal CtrB into the cytoplasm as a result of lysosomal membrane permeabilization (LMP). Suppression of CtrB significantly blocked apoptosis, while overexpression of CtrB sensitized apoptosis markedly. CtrB could cleave Bid under neutral conditions. In RH-35 cells with Bid silenced, apoptosis induced by CtrB protein was attenuated, suggesting that CtrB mediates apoptosis of RH-35 cells mainly through processing Bid. Our data also suggest that LMP occurs earlier than mitochondrial outer membrane permeabilization; Bid activation initiated by caspase-8 might be reinforced by CtrB in consequence of LMP, which causes a positive feedback loop leading to the accumulation of tBid, and results in lysosome- and mitochondrion-dependent apoptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0554
4-Acetamido-4′-isothiocyanato-2,2′-stilbenedisulfonic acid disodium salt hydrate, ≥80%
C17H12N2Na2O7S3 · xH2O