EMAIL THIS PAGE TO A FRIEND
Endocrinology
Sertoli Cell-Initiated Testicular Innate Immune Response through Toll-Like Receptor-3 Activation Is Negatively Regulated by Tyro3, Axl, and Mer Receptors.
PMID 20363878