EMAIL THIS PAGE TO A FRIEND
Physical review letters
Diffraction properties of periodic lattices under free electron laser radiation.
PMID 20366545
Abstract
In this Letter, we report the pioneering use of free electron laser radiation for the investigation of periodic crystalline structures. The diffraction properties of silver behenate single nanocrystals (5.8 nm periodicity) with the dimensions of 20 nm x 20 nm x 20 microm and as powder with grain sizes smaller than 200 nm were investigated with 8 nm free electron laser radiation in single-shot modus with 30 fs long free electron laser pulses. This work emphasizes the possibility of using soft x-ray free electron laser radiation for these crystallographic studies on a nanometer scale.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11909
Behenic acid, analytical standard
C22H44O2
216941
Behenic acid, 99%
C22H44O2