EMAIL THIS PAGE TO A FRIEND
Acta Obstetricia et Gynecologica Scandinavica
The effect of nicotine on the production of soluble fms-like tyrosine kinase-1 and soluble endoglin in human umbilical vein endothelial cells and trophoblasts.
PMID 20367431