EMAIL THIS PAGE TO A FRIEND
Cancer research and treatment : official journal of Korean Cancer Association
Prognostic significance of immunohistochemical expression of EGFR and C-erbB-2 oncoprotein in curatively resected gastric cancer.
PMID 20368841
Abstract
The purpose of this study was to investigate the prognostic significance of the expression of EGFR and C-erbB-2 gene products by immunohistochemical analysis for curatively resected gastric adenocarcinoma. Between January 1996 and December 2001, 739 patients with curatively resected gastric cancer patients underwent immunohistochemical staining for EGFR and C-erbB-2 proteins, and we retrospectively analyzed their correlation with the clinical outcome. The over expressions of EGFR and C-erbB-2 were 25.4% and 26.2%, respectively. The over expressions of EGFR was associated with the more poorly differentiated tumor (p=0.000) and with neuronal invasion (p=0.03). Over expression of C-erbB-2 was associated with less vascular invasion (p=0.001). Tumor depth or node metastasis was not related to the over expression of EGFR or C-erbB-2. The seven-year overall survival and relapse-free survival rates were 87.2% and 75.8%, respectively. Upon multivariate Cox regression analysis, the tumor stage, tumor size and patient age were important prognostic factors for overall survival, and tumor stage was the important factor for relapse-free survival. Over expressions of EGFR or c-erbB-2 were not significant prognostic factors. Immunohistochemical staining of EGFR and C-erbB-2 gene products were not independent prognostic factors for predicting the overall survival and the relapse-free survival in curatively resected gastric cancer.