EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Novel cellulose-based polyelectrolytes synthesized via the click reaction.
PMID 20371038
Abstract
Novel polyelectrolytes were prepared by conversion of 6-azido-6-deoxycellulose with acetylenedicarboxylic acid dimethyl ester and subsequent saponification. Up to 62% of the azide moieties were converted. The reaction was completed within 48 h using 2 moles of acetylenedicarboxylic acid dimethyl ester per mole of modified anhydroglucose unit. FTIR and NMR spectroscopy were applied to elucidate the molecular structure of the polymers. The polymer degradation was acceptable during this two-step reaction. The resulting biopolymer derivatives were water soluble and reduced the surface tension on water significantly. Moreover, they form ionotropic gels with multivalent metal ions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D138401
Dimethyl acetylenedicarboxylate, 99%
C6H6O4