EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Chemical and in vitro study of the potential of 3-(aryl)-2-sulfanylpropenoic acids and their Zn(II) complexes as protective agents against cadmium toxicity.
PMID 20372718
Abstract
The free H(2)xspa ligands [xspa = pspa, Clpspa, tspa or fspa where p = 3-(phenyl), Clp = 3-(2-chlorophenyl), t = 3-(2-thienyl), f = 3-(2-furyl) and spa = 2-sulfanylpropenoato], their Zn(II) complexes of formula [HQ](2)[Zn(xspa)(2)] (HQ = diisopropylammonium) and the Cd(II) equivalents were prepared and characterized by elemental analysis and by IR, Raman and NMR ((1)H, (13)C) spectroscopy. X-Ray studies of the crystal structures of [HQ](2)[Zn(pspa)(2)], [HQ](2)[Zn(Clpspa)(2)], [HQ](2)[Zn(tspa)(2)] and [HQ](2)[Zn(fspa)(2)] show that the zinc atom is coordinated to two O atoms and two S atoms of the ligands in a distorted tetrahedral ZnO(2)S(2) environment. In the structures of [HQ](2)[Cd(pspa)(2)] and [HQ](2)[Cd(Clpspa)(2)] the cadmium atom is coordinated to three S atoms and two carboxylato O atoms of the ligands in a distorted trigonal bipyramidal environment. The interchange of ligands between Zn(II) and Cd(II) was studied by (113)Cd NMR spectroscopy. The in vitro protective effect of H(2)xspa and their Zn(II) complexes against Cd toxicity was investigated using the human hepatocarcinoma HepG2 cell line and the pig renal proximal tubule LLC-PK1 cell line. The incorporation of Zn(II) was found to be relevant in the case of H(2)pspa, with an increase observed in the cell viability of the LCC-PK1 cells with respect to the value for the free ligand.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
LLC-PK1, 86121112