EMAIL THIS PAGE TO A FRIEND
Molecular diversity
An efficient, multicomponent approach for solvent-free synthesis of 2-amino-4H-chromene scaffold.
PMID 20373141
Abstract
Solvent-free one-pot synthesis of 2-amino-4H-chromene scaffold is described in a very simple, efficient, and environmentally benign method using sodium carbonate as a cheap and non-toxic catalyst with up to excellent yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
112216
4-Chlorobenzaldehyde, 97%
C7H5ClO