EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Antimicrobial activity of tryptanthrins in Escherichia coli.
PMID 20373766
Abstract
Tryptanthrins have potential therapeutic activity against a wide variety of pathogenic organisms, although little is known about their mechanism. Activity against Escherichia coli, however, has not been examined. The effects of tryptanthrin (indolo[2,1-b]quinazolin-6,12-dione) and nine derivatives on growth, survival, and mutagenesis in E. coli were examined. Analogues with a nitrogen atom at the 4-position of tryptanthrin stopped log phase growth of E. coli cultures at concentrations as low as 5 microM. Tryptanthrins decreased viability during incubation with cells in buffer by factors of 10(-2) to <10(-6) at 0.2-40 microM. Derivatives with an oxime group at the 6-position exhibited the greatest bactericidal activity. Most tryptanthrins were not mutagenic in several independent assays, although the 4-aza and 4 aza-8-fluoro derivatives increased frameshift mutations about 22- and 4-fold, respectively. Given the structure of trypanthrins, binding to DNA may occur by intercalation. From analysis using a sensitive linking number assay, several tryptanthrins, especially the 4-aza and 6-oximo derivatives, intercalate into DNA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0310
Tryptanthrin, ≥98% (HPLC)
C15H8N2O2