EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Effect of substituents on the excited-state dynamics of the modified DNA bases 2,4-diaminopyrimidine and 2,6-diaminopurine.
PMID 20379571
Abstract
To explore the excited state dynamics of pyrimidine derivatives, we performed a combined experimental and theoretical study. We present resonant two-photon ionization (R2PI) and IR-UV double resonance spectra of 2,4-diaminopyrimidine and 2,6-diaminopurine seeded in a supersonic jet by laser desorption. For 2,4-diaminopyrimidine (S(0)-->S(1) 34,459 cm(-1)), we observed only the diamino tautomer with an excited state lifetime bracketed between experimental limits of 10 ps and 1 ns. For 2,6-diaminopurine, we observed two tautomers, the 9H- (S(0)-->S(1) 34,881 cm(-1)) and 7H- (S(0)-->S(1) 32,215 cm(-1)) diamino forms, with excited state lifetimes of 6.3±0.4 ns and 8.7±0.8 ns, respectively. We investigated the nature of the excited state of 2,4-diaminopyrimidine by means of multi-reference ab initio methods. The calculations of stationary points in the ground and excited states, minima on the S(0)/S(1) crossing seam and connecting reaction paths show that several paths with negligible barriers exist, allowing ultrafast radiationless deactivation if excited at energies slightly higher than the band origin. The sub-nanosecond lifetime found experimentally is in good agreement with this finding.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
468231
2,4-Diaminopyrimidine, 98%
C4H6N4
247847
2,6-Diaminopurine, 98%
C5H6N6