EMAIL THIS PAGE TO A FRIEND
Mini reviews in medicinal chemistry
Gastrin: an acid-releasing, proliferative and immunomodulatory peptide?
PMID 20380639
Abstract
Gastrin release is affected by gastric inflammatory conditions. Antral G cells respond to inflammatory mediators by increasing gastrin secretion. Accumulating experimental evidence suggests that gastrin exerts immunomodulatory and proinflammatory effects. Gastrin could be a contributing factor to these pathologies, which may constitute a new justification for pharmacological blockade of gastrin action.
Related Materials