EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Transkarbams as transdermal permeation enhancers: effects of ester position and ammonium carbamate formation.
PMID 20381349
Abstract
Transkarbam 12, an ammonium carbamate formed by the reaction of dodecyl 6-aminohexanoate with carbon dioxide, is a highly active, broad-spectrum, nontoxic, and nonirritant transdermal permeation enhancer. It probably acts by a dual mechanism: a part of its activity is associated with the carbamic acid salt and/or its decomposition in the acidic stratum corneum. The ammonium ester thereby released is an active enhancer species as well, and its activity highly depends on the position of the ester group.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09699
Ammonium carbamate, for decomposition
CH6N2O2
292834
Ammonium carbamate, 99%
CH6N2O2