EMAIL THIS PAGE TO A FRIEND
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie
Erythrocytes oxidative damage and hematological effects of 2,4,4',5-tetrachlorodiphenyl sulfone in rats.
PMID 20382005
Abstract
The effects of subchronic exposure to tetrachlorodiphenyl sulfone (TCDS) on hematological parameters [white blood cells (WBC), red blood cells (RBC), mean cell volume (MCV), mean corpuscular hemoglobin (MCH), mean corpuscular hemoglobin concentration (MCHC), hemoglobin (Hb) and hematocrite (Ht) levels] were examined. Oxidative stress in erythrocytes was also assessed by measuring thiobarbituric acid reactive substances (TBARS) and enzyme antioxidant activities [superoxide dismutase (SOD), catalase (CAT), and glutathione peroxidase (GPx)]. TCDS was administered orally, dissolved in water, ad libitum to 12 female rats at 28.9 mg/kg/day for 6 or 12 weeks. Results showed that TCDS induced significant decreases in RBC, Hb, and Ht. Whereas MCV, MCH, and MCHC remain unchanged and WBC increased only in the second period of the study. Moreover erythrocyte TBARS level increased, and antioxidant enzyme (SOD, GPx, and CAT) activities decreased. We concluded that TCDS intoxication promotes erythrocyte oxidative damage and disrupt hematological constituents in rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45680
Tetradifon, PESTANAL®, analytical standard
C12H6Cl4O2S