EMAIL THIS PAGE TO A FRIEND
Nature methods
Using buoyant mass to measure the growth of single cells.
PMID 20383132
Abstract
We used a suspended microchannel resonator (SMR) combined with picoliter-scale microfluidic control to measure buoyant mass and determine the 'instantaneous' growth rates of individual cells. The SMR measures mass with femtogram precision, allowing rapid determination of the growth rate in a fraction of a complete cell cycle. We found that for individual cells of Bacillus subtilis, Escherichia coli, Saccharomyces cerevisiae and mouse lymphoblasts, heavier cells grew faster than lighter cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L3397 LB Broth (Luria low salt), Powder microbial growth medium