EMAIL THIS PAGE TO A FRIEND
The European Journal of Neuroscience
Neocortical synaptophysin asymmetry and behavioral lateralization in chimpanzees (Pan troglodytes).
PMID 20384782
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB3418
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse