EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Six-component reactions for the stereoselective synthesis of 3-arylidene-2-oxindoles via sequential one-pot Ugi/Heck carbocyclization/Sonogashira/nucleophilic addition.
PMID 20387848
Abstract
An efficient palladium-catalyzed protocol for the synthesis of 3-arylidene-2-oxindoles has been developed. In this approach, a sequential one-pot six-component reaction via Ugi/Heck carbocyclization/Sonogashira/nucleophilic addition was used for the synthesis of the desired skeleton.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O9808
2-Oxindole, 97%
C8H7NO