EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Efficient syntheses of clofarabine and gemcitabine from 2-deoxyribonolactone.
PMID 20391198
Abstract
The development of a new methodology to achieve electrophilic fluorination of triisopropylsilyl-protected 2-deoxyribonolactone has been employed to synthesize clofarabine and gemcitabine with improved synthetic efficiency versus prior synthetic methods. These studies highlight the versatility of this new methodology to obtain medically relevant 2'-fluoronucleosides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30957
2-Deoxy-D-ribono-1,4-lactone, ≥95% (GC)
C5H8O4