EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A tumor-acidity-activated charge-conversional nanogel as an intelligent vehicle for promoted tumoral-cell uptake and drug delivery.
PMID 20391548
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D167800
2,3-Dimethylmaleic anhydride, 98%
C6H6O3