EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Rare oxidation-state combinations and unusual structural motifs in hexanuclear Mn complexes using 2-pyridyloximate ligands.
PMID 20392125
Abstract
The use of phenyl-2-pyridyl ketone oxime and di-2-pyridyl ketone oxime in Mn chemistry has led to hexanuclear clusters with unprecedented (Mn(II)(4)Mn(III)Mn(IV)) or extremely rare (Mn(II)Mn(III)(5) and Mn(II)(3)Mn(III)(3)) metal oxidation-state combinations and uncommon structural motifs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161705
Di-2-pyridyl ketone oxime, 98%
C11H9N3O