EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Choline promotes nicotinic receptor alpha4 + beta2 up-regulation.
PMID 20392695
Abstract
Neuronal nicotinic acetylcholine receptors (nAChR) composed of alpha4 + beta2 subunits, the high affinity nicotine-binding site in the mammalian brain, up-regulate in response to chronic nicotine exposure. The identities of endogenous mediators of this process are unknown. We find that choline also up-regulates alpha4 + beta2 nAChRs stably expressed by HEK293 cells as measured by increased [(3)H]epibatidine density. Choline-mediated up-regulation is dose-dependent and corresponds with an increase in beta2 subunit protein expression. The choline kinase inhibitor hemicholinium-3 inhibits approximately 60% of choline-mediated up-regulation revealing both an HC3-dependent and -independent pathway. Furthermore, choline-mediated up-regulation is not additive with up-regulation agents such as nicotine, but it is additive with weaker promoters of the up-regulation process. When co-applied with the pro-inflammatory cytokine tumor necrosis factor alpha, choline-mediated up-regulation is increased further through a mechanism that includes an increase in both alpha4 and beta2 protein expression, and this is inhibited by the p38 MAPK inhibitor SB202190. These findings extend the view that up-regulation of alpha4 + beta2 nAChRs is a normal physiological response to altered metabolic and inflammatory conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H108
Hemicholinium-3, solid, ≥95% (HPLC)
C24H34Br2N2O4