EMAIL THIS PAGE TO A FRIEND
Marine biotechnology (New York, N.Y.)
Microarray analyses of shrimp immune responses.
PMID 20393773
Abstract
Shrimp aquaculture is one of the major foodproducing industries in the world. However, it is being impacted by several problems including diseases, antibiotic use, and environmental factors. The extent of the effects of these problems in the immune system of the shrimp at the molecular level is just beginning to be understood. Here, we review the gene expression profile of shrimp in response to some of these problems using the high-throughput microarray analysis, including white spot syndrome virus, yellow head virus, Vibrio spp., peptidoglycan, oxytetracycline, oxolinic acid, salinity, and temperature.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O0877
Oxolinic acid, quinolone antibiotic
C13H11NO5
67126
Oxolinic acid, analytical standard
C13H11NO5