Langmuir : the ACS journal of surfaces and colloids
Magnetic alloy nanoparticles from laser ablation in cyclopentanone and their embedding into a photoresist.
PMID 20394393
The generation of nonoxidized magnetic alloy nanoparticles is still a challenge using conventional chemical reduction methods. However, because these nanoparticles are currently attracting much attention, alternative methods are required. In this context, the applicability of femtosecond laser ablation, which has evolved as a powerful tool for the generation of colloidal metal nanoparticles, has been investigated using the example of Ni(48)Fe(52) and Sm(2)Co(17) ablation in cyclopentanone. Besides stability and size measurements, the focus has been placed on the analysis of the elemental composition of nanoparticles, which proved the preservation of the stoichiometry of the target in Ni-Fe nanoparticles but not in Sm-Co. It is assumed that this is due to a greater difference in the heat of evaporation of the bulk alloy components in Sm-Co than in Ni-Fe. Hence, the successful generation of magnetic alloy nanoparticles is possible for alloys composed of elements with similar heats of evaporation. This one-step approach allows the fabrication of nanomagnetic polymer composites (e.g., with application prospects in microtechnology such as microactuators).