EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
The synthesis of novel fluorescent purine analogues modified by azacrown ether at C6.
PMID 20399655
Abstract
The synthesis and fluorescence properties of novel purine analogues linked azacrown ether at C6 position were investigated. These new purine analogues could be prepared from a series of 6-chloropurines and showed selective and efficient signaling behaviors toward micromolar concentration of Ag(+) ion over other common metal ions in an aqueous environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161179
6-Chloropurine, ≥99%
C5H3ClN4
511617
6-Chloropurine, 99%
C5H3ClN4
26260
6-Chloropurine, ≥95.0% (HPLC)
C5H3ClN4