EMAIL THIS PAGE TO A FRIEND
Chemosphere
Effect of size-fractionation dissolved organic matter on the mobility of prometryne in soil.
PMID 20400172
Abstract
Import of organic materials in the form of compost, sludge or plant residues introduces large amounts of dissolved organic matter (DOM) into soils. DOM as a dynamic soil component affects the behaviors of organic pollutants. Different DOM constituents may affect herbicide action in a different way. However, the process of interaction between the distinct DOM-fractions and herbicides is largely unknown. In this study, DOM was separated by size-fractionation into three molecular size groups: MW<3500 Da, 3500 Da14000 Da. Effects of DOM-fractions on prometryne sorption/desorption and mobility were analyzed using approaches of batch experiments, soil column and soil thin-layer chromatography. Application of varied DOM-fractions at 50mg DOCL(-1) to the soil reduced the sorption and increased desorption of prometryne. DOM-fraction with MW>14000 Da appeared most effective in prometryne mobilization in the soil than any other fractions. Finally, DOM-fractions were characterized by chemical analyses, fourier transformed infrared spectroscopy (FT-IR) and excitation-emission matrices (EEMs) fluorescence spectroscopy. Our studies revealed that the high-molecular weight fraction contained more aromatic framework and unsaturated structure that was most likely the dominant factor modulating the behavior of prometryne in soils.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45636
Prometryn, PESTANAL®, analytical standard
C10H19N5S