EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Design and synthesis of novel SATE derivatives of acyclic isocytosine and 9-deazaadenine C-nucleosides.
PMID 20408056
Abstract
This article describes a very simple route for synthesizing novel lipophilic phosphate bis(t-bu-SATE) prodrugs of acyclic cyclobutylated C-nucleosides such as isocytosine 12 and 9-deazaadenine 19, which were prepared from 1,1-gem cyclobutyl dicarboxylate. Synthesized compounds were evaluated as potential antiviral agents against HIV virus. Some phosphate SATE prodrugs were more active against HIV than parent nucleosides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2127
Isocytosine, ≥99%
C4H5N3O
T0642
Tubercidin, from Streptomyces tubercidicus, ~95%
C11H14N4O4