EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Comparative study of new shell-type, sub-2 micron fully porous and monolith stationary phases, focusing on mass-transfer resistance.
PMID 20409553
Abstract
Today sub-2 microm packed columns are very popular to conduct fast chromatographic separations. The mass-transfer resistance depends on the particle size but some practical limits exist not to reach the theoretically expected plate height and mass-transfer resistance. Another approach applies particles with shortened diffusion path to enhance the efficiency of separations. In this study a systematical evaluation of the possibilities of the separations obtained with 5 cm long narrow bore columns packed with new 2.6 microm shell particles (1.9 microm nonporous core surrounded by a 0.35 microm porous shell, Kinetex, Core-Shell), packed with other shell-type particles (Ascentis Express, Fused-Core), totally porous sub-2 microm particles and a 5 cm long narrow bore monolith column is presented. The different commercially available columns were compared by using van Deemter, Knox and kinetic plots. Theoretical Poppe plots were constructed for each column to compare their kinetic performance. Data are presented on polar neutral real-life analytes. Comparison of a low molecular weight compounds (MW=270-430) and a high molecular weight one (MW approximately 900) was conducted. This study proves that the Kinetex column packed with 2.6 microm shell particles is worthy of rivaling to sub-2 microm columns and other commercially available shell-type packings (Ascentis Express or Halo), both for small and large molecule separation. The Kinetex column offers a very flat C term. Utilizing this feature, high flow rates can be applied to accomplish very fast separations without significant loss in efficiency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51594-U Ascentis® Express RP-Amide, 2 Micron Guard Cartridge, 2 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
51595-U Ascentis® Express RP-Amide, 2 Micron Guard Cartridge, 2 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
51567-U Ascentis® Express RP-Amide, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 2 cm × 2.1 mm
51568-U Ascentis® Express RP-Amide, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 3 cm × 2.1 mm
51569-U Ascentis® Express RP-Amide, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 2.1 mm
51571-U Ascentis® Express RP-Amide, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 7.5 cm × 2.1 mm
51576-U Ascentis® Express RP-Amide, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 2.1 mm
51577-U Ascentis® Express RP-Amide, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 15 cm × 2.1 mm
51582-U Ascentis® Express RP-Amide, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 3 cm × 3 mm
51583-U Ascentis® Express RP-Amide, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 3 mm
51587-U Ascentis® Express RP-Amide, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 7.5 cm × 3 mm
51588-U Ascentis® Express RP-Amide, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 3 mm
51589-U Ascentis® Express RP-Amide, 2 Micron UHPLC Column, 2 μm particle size, L × I.D. 15 cm × 3 mm
53514-U Ascentis® Express RP-Amide, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
53516-U Ascentis® Express RP-Amide, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
53519-U Ascentis® Express RP-Amide, 2.7 Micron Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea
53910-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 2.1 mm
53911-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 2.1 mm
53912-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 7.5 cm × 2.1 mm
53913-U
Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
53914-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 2.1 mm
53915-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 3 mm
53916-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 3 mm
53917-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 7.5 cm × 3 mm
53918-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 10 cm × 3 mm
53919-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 3 mm
53921-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 3 cm × 4.6 mm
53922-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 4.6 mm
53923-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 7.5 cm × 4.6 mm
53929-U Ascentis® Express RP-Amide, 2.7 Micron HPLC Column, 2.7 μm particle size, L × I.D. 10 cm × 4.6 mm