EMAIL THIS PAGE TO A FRIEND
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society
Integrin alpha3 subunit regulates events linked to epithelial repair, including keratinocyte migration and protein expression.
PMID 20412552
Abstract
Two integrins, alpha3beta1 and alpha6beta4, are high-affinity receptors for laminin 332, the major laminin isoform of the dermal-epidermal junction, although they are thought to have different functions. Biological and genetic studies have firmly established that the alpha6beta4 integrin is indispensable for the stable anchorage of the epidermis to the underlying dermis. In contrast, the alpha3beta1 integrin is thought to be important for cell migration, although the issue is controversial, and both positive and negative effects have been reported. To address the function of alpha3beta1 integrin, we used small interfering RNA to down-regulate the alpha3 subunit in human keratinocytes. The resulting phenotype indicates that lack of alpha3beta1 integrin compromises intercellular adhesion and collective migration, while it enhances single cell migration with a concomitant increase of both focal adhesion kinase and extracellular signal-regulated kinase. In addition, down-regulation of integrin alpha3 subunit results in an increased expression of fibronectin and precursor laminin 332, two extracellular matrix proteins known to be up-regulated during wound healing. Thus, down-regulation of alpha3beta1 integrin recapitulates crucial events governing keratinocyte migration associated with wound healing and tissue repair.