EMAIL THIS PAGE TO A FRIEND
Obesity (Silver Spring, Md.)
Body composition and energy metabolism following Roux-en-Y gastric bypass surgery.
PMID 20414197
Abstract
Roux-en-Y gastric bypass (RYGB) surgery has become an accepted treatment for excessive obesity. We conducted a longitudinal study to assess regional body composition, muscle proteolysis, and energy expenditure before RYGB, and 6 and 12 months after RYGB. Whole-body and regional fat mass (FM) and lean mass (LM) were assessed via dual energy X-ray absorptiometry (DXA), and myofibrillar protein degradation was estimated by urinary 3-methylhistidine (3-MeH) in 29 subjects. Energy expenditure and substrate oxidation were also determined using a whole-room, indirect calorimeter in 12 of these subjects. LM loss constituted 27.8 +/- 10.2% of total weight loss achieved 12 months postoperatively, with the majority of LM loss (18 +/- 6% of initial LM) occurring in the first 6 months following RYGB. During this period, the trunk region contributed 66% of whole-body LM loss. LM loss occurred in the first 6 months after RYGB despite decreased muscle protein breakdown, as indicated by a decrease in 3-MeH concentrations and muscle fractional breakdown rates. Sleep energy expenditure (SEE) decreased from 2,092 +/- 342 kcal/d at baseline to 1,495 +/- 190 kcal/day at 6 months after RYGB (P < 0.0001). Changes in both LM and FM had an effect on the reduction in SEE (P < 0.001 and P = 0.005, respectively). These studies suggest that loss of LM after RYGB is significant and strategies to maintain LM after surgery should be explored.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M9005
3-Methyl-L-histidine
C7H11N3O2