EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Isolation and characterization of 23-O-mycinosyl-20-dihydro-rosamicin: a new rosamicin analogue derived from engineered Micromonospora rosaria.
PMID 20414322
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R2636
Rosamicin, from Micromonospora rosaria, ~98%
C31H51NO9