EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Nitroxide polymer networks formed by Michael addition: on site-cured electrode-active organic coating.
PMID 20414502
Abstract
Highly and homogeneously crosslinked poly(beta-ketoester) networks densely bearing robust nitroxide radicals were prepared via a click-type and stepwise Michael polyaddition. A half-battery cell composed of the thermally-cured radical network coatings displayed a rapid, reversible, and almost stoichiometric redox-activity even with a thickness of ca. 10 mum, which may be applicable as the electrode of organic-based rechargeable devices.