Zhongguo gu shang = China journal of orthopaedics and traumatology
[Experimental study on avascular necrosis of femoral head in chickens induced by different glucocorticoides].
PMID 20415073
To study the effects of Methylprednisolone and Dexamethasone on the avascular necrosis of femoral head in chickens. Thirty-six chickens were randomly divided into 6 groups (n = 6): control group (group A), Methylprednisolone low dose group (group B), Methylprednisolone large dose group (group C), small dose Dexamethasone and horse serum group (group D), middle dose Dexamethasone and horse serum group (group E), and Dexamethasone large dose group (group F). On the 6th and 12th weeks, blood samples were obtained to determine the level of total cholesterol triglyeride (TG), high density lipoprotein (HDL) and low density lipoprotein (LDL). On the 12th week, femoral heads were taken off. Paraffin tissue sections were prepared to detect histopathologic change with hematoxylin and eosin staining. On the 6th week, compared with group A, the level of CHO increased significantly in group C and group F (P < 0.05), and TG increased in group B, C and group E, while HDL decreased in group B, C and group E. On the 12th week, the level of TG and CHO increased in group B, C, E and group F, and HDL decreased in group C, D and group E (P < 0.05). LDL was not detected in most chickens. The ratio of empty lacuna was higher in group C and group E compared with those of the control group (P < 0.05). Methylprednisolone is easier to induce osteonecrosis of femoral head than Dexamethasone. The condition of metabolic disorder in blood may be the basic pathomechanism of steroid-induced necrosis of femoral head.
Related Materials
Product #
Molecular Formula
Add to Cart
Gly-Gly-Gly, BioUltra, ≥99.0% (NT)