EMAIL THIS PAGE TO A FRIEND
Alimentary pharmacology & therapeutics
Alverine citrate, simeticone, and Rome III irritable bowel syndrome.
PMID 20415841
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0424
Alverine citrate salt
C20H27N · C6H8O7
Y0000687
Alverine citrate salt, European Pharmacopoeia (EP) Reference Standard
C20H27N · C6H8O7