EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A one-step synthesis of 2,4-unsubstituted quinoline-3-carboxylic acid esters from o-nitrobenzaldehydes.
PMID 20420395
Abstract
A straightforward and efficient one-step procedure for the synthesis of 2,4-unsubstituted quinoline-3-carboxylic acid ethyl esters is described. The simple reductive cyclization is carried out by treating various substituted o-nitrobenzaldehydes with inexpensive, commercially available 3,3-diethoxypropionic acid ethyl ester and SnCl(2).2H(2)O in refluxing ethanol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N10802
2-Nitrobenzaldehyde, 98%
C7H5NO3