EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Reactivity of the insecticide fenitrothion toward O and N nucleophiles.
PMID 20423065
Abstract
The reactivity of Fenitrothion (1) toward several O- and N-based nucleophiles, including ambident and alpha-nucleophiles, was investigated in basic media at 25 degrees C in water containing 2% 1,4-dioxane. In the reactions with HO(-) and HOO(-) quantitative formation of 3-methyl-4-nitrophenoxide (2) was observed indicating a S(N)2(P) pathway. In the reactions with NH(2)OH, NH(2)O(-), and BuNH(2), demethylfenitrothion (4) was formed along with 2, indicating competition between the S(N)2(P) and S(N)2(C) pathways; no evidence of a S(N)Ar pathway was observed in any case. The observed rate constants were dissected into the values corresponding to the S(N)2(P) and S(N)2(C) pathways. The yield of 4 depends on the nucleophile and on the pH of the reaction, being the main product in the case of BuNH(2). With HOO(-), NH(2)OH, and NH(2)O(-) a significant alpha-effect was observed, confirming the participation of the nucleophile in the rate-limiting step of the reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W313009
Butylamine, ≥99%
C4H11N
471305
Butylamine, 99.5%
C4H11N