EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Palladium-catalyzed domino C-C/C-N coupling using a norbornene template: synthesis of substituted benzomorpholines, phenoxazines, and dihydrodibenzoxazepines.
PMID 20423091
Abstract
A rapid, four-step approach to alkyl- and aryl-substituted benzomorpholines is accomplished by a Pd-catalyzed domino C-C/C-N bond coupling using a norbornene template. Extension to phenoxazines and dihydrodibenzoxazepines is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
263893
Phenoxazine, ≥99%, purified by sublimation
C12H9NO
P14858
Phenoxazine, 97%
C12H9NO