EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Selective Heck arylation of cyclohexene with homogeneous and heterogeneous palladium catalysts.
PMID 20428035
Abstract
Palladium catalysts containing Pd(II) supported on Al2O3 and alumina-based mixed oxides, Al2O3-ZrO2, Al2O3-CeO2, and Al2O3-Fe2O3, are very effective in the Heck coupling of iodobenzene with cyclohexene in DMF solution. The best results, up to 81% of monoarylated products with a selectivity to 4-phenylcyclohexene (3) close to 90% were obtained with KOH as a base. The catalytic activity of palladium supported on alumina-based oxides was compared with that of homogeneous precursors, such as Pd(OAc)2 and PdCl2(PhCN)2, used in [Bu4N]Br as the reaction medium. Under such conditions homogeneous systems were more selective and produced up to 60% of monoarylated products with a selectivity to 3 close to 60%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7632
Iodobenzene, 98%
C6H5I