EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Synthesis and antimicrobial activity of new 4-heteroarylamino coumarin derivatives containing nitrogen and sulfur as heteroatoms.
PMID 20428040
Abstract
Synthesis, spectral analysis and bioactivity of new coumarin derivatives are described in this paper. Eight new coumarin derivatives were synthesized in moderate to good yields by condensation of 4-chloro-3-nitrocoumarin and the corresponding heteroarylamine. The synthesized compounds were tested for their in vitro antimicrobial activity, in a standard disk diffusion assay, against thirteen strains of bacteria and three fungal strains. They have shown a wide range of activity - from one completely inactive compound to medium active ones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
438626
4-Hydroxy-3-nitrocoumarin, 98%
C9H5NO5