EMAIL THIS PAGE TO A FRIEND
Organic letters
Chiral Brønsted acid catalyzed enantioselective addition of alpha-isocyanoacetamides to aldehydes.
PMID 20429535
Abstract
A clean and highly efficient enantioselective addition of alpha-isocyanoacetamides to aliphatic aldehydes catalyzed by chiral phosphoric acid in the presence of MS 5 A in toluene at -40 degrees C was developed. Excellent yields (85-98%) and good to excellent enantioselectivities (up to >99% ee) were achieved.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
108448
Cyanoacetamide, 99%
C3H4N2O